Learn More
Chemglass Life Sciences Complete: 6mm Bore, 16/35 Plug Size, 11 & 12mm OD Sidearms
SDP
Catalog No. 501531999
Encompass_Preferred
Complete: 6mm Bore, 16/35 Plug Size, 11 & 12mm OD Sidearms. Similar to CG-424 but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Available complete with 1:5 taper PTFE plug or glass shell only. FOR REPLACEMENT PLUGS, SEE CG-424 LISTING
Catalog No. 50-153-1999 Supplier Chemglass Life Sciences Supplier No. CGB42803
Complete: 6mm Bore, 16/35 Plug Size, 11 & 12mm OD Sidearms. Similar to CG-424 but with one arm of the stopcock being of a larger diameter. Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Available complete with 1:5 taper PTFE plug or glass shell only. FOR REPLACEMENT PLUGS, SEE CG-424 LISTING