Learn More
Ace Glass, Inc. 6MM STOPCOCK GB 6MM STOPCOCK GB
SDP
Catalog No. 50976118
Encompass_Preferred
Glass 6mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.
Catalog No. 50-976-118 Supplier Ace Glass, Inc. Supplier No. 813710
Glass 6mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.