Learn More
AVANTOR MASTERFLEX (VWR LLC) Fitting, PFA, Straight, Compression Union, 6mm OD
Catalog No. NC9461224
Encompass
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. NC9461224 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3132164
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.