Learn More
MSC Norgren C02250628 6mm Tube OD, 1/4 BSPP Brass Push-to-Connect Tube Male Connector Tube to Male BSPP Connection, 150 Max psi, Nitrile O-Ring, 0 to 175°F Working Temp
Catalog No. NC1683007
Encompass
Norgren C02250628 6mm Tube OD, 1/4 BSPP Brass Push-to-Connect Tube Male Connector Tube to Male BSPP Connection, 150 Max psi, Nitrile O-Ring, 0 to 175°F Working Temp
Catalog No. NC1683007 Supplier MSC Supplier No. 00350660
Norgren C02250628 6mm Tube OD, 1/4 BSPP Brass Push-to-Connect Tube Male Connector Tube to Male BSPP Connection, 150 Max psi, Nitrile O-Ring, 0 to 175°F Working Temp