Learn More
Ace Glass, Inc. PTFE 6mm tube compression fitting to 1/4in male NPT
SDP
Supplier:  Ace Glass, Inc. 1270926
Encompass
6mm PTFE tube compression fitting to 1/4 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-677