Learn More
Ace Glass, Inc. PTFE 6mm tube compression fitting to 1/8in male NPT
SDP
Catalog No. 50983396
Encompass
6mm PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-983-396 Supplier Ace Glass, Inc. Supplier No. 1270914
6mm PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.