Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6N Hydrochloric Acid Aqueous Solution, Cenmed™
SDP

Catalog No. 21520250
Change view
Click to view available options
Quantity:
4 x 60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-250 4 x 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-250 Supplier Cen-Med Enterprises Supplier No. CENMED 3250202
Add to Cart
Add to Cart

Unlock versatility in your lab experiments with 6N Hydrochloric Acid Aqueous Solution, a lab-grade reagent meticulously formulated for controlled experiments, providing a reliable and precise solution for various applications.

  • Packaged in 60 mL plastic vials for easy handling and storage, providing convenience and minimizing the risk of spills or contamination
  • 6N concentration ensures optimal performance, meeting the stringent requirements of scientific research and delivering consistent results
  • Stand out from the rest with a lab reagent that combines quality, convenience, and safety, catering to the needs of researchers and scientists
  • Ensure precision in your experiments by using a solution that minimizes the risk of errors, enhancing the overall reliability of your results
  • Researchers can confidently explore various applications with a versatile solution designed to meet the demands of diverse laboratory scenarios

Specifications

Physical Form Aqueous Solution, Liquid
CAS 7647-01-0
Packaging Plastic Vial
Color Clear
Quantity 4 x 60 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.