Learn More
Hach Company Duraprene pump tubing, 0.040 in (1.016 mm) I.D., white/white, pack of 6 - replacement tubing for peristaltic and metering pumps
Catalog No. NC2018526 Shop All Hach Company Products
Encompass
Duraprene pump tubing with a 0.040 in (1.016 mm) inner diameter, white on both surfaces, sold as a package of six. Designed for use as replacement or spare tubing in peristaltic and metering pumps, it provides consistent flow and compatibility for liquid handling and sampling applications. Ships with manufacturer lead time as noted on the product page.
  • Duraprene material for flexibility and pump compatibility.
  • 0.040 in (1.016 mm) inner diameter for controlled flow.
  • White/white color for easy visual inspection of fluid movement.
  • Pack of six tubes to support maintenance and spare needs.
  • Suitable for peristaltic and metering pump applications.
Catalog No. NC2018526 Supplier Hach Company Supplier No. 54409
Duraprene pump tubing with a 0.040 in (1.016 mm) inner diameter, white on both surfaces, sold as a package of six. Designed for use as replacement or spare tubing in peristaltic and metering pumps, it provides consistent flow and compatibility for liquid handling and sampling applications. Ships with manufacturer lead time as noted on the product page.
  • Duraprene material for flexibility and pump compatibility.
  • 0.040 in (1.016 mm) inner diameter for controlled flow.
  • White/white color for easy visual inspection of fluid movement.
  • Pack of six tubes to support maintenance and spare needs.
  • Suitable for peristaltic and metering pump applications.