Learn More
Chemglass Life Sciences Collection Vial Rack Only, 6-Place, Bill-Board
SDP
Catalog No. 501531269
Encompass_Preferred
Collection Vial Rack Only, 6-Place, Bill-Board. Have six wells to hold 25 x 52mm collection vials. Racks have four Stainless Steel pins that correspond to the holes in the top plate assemblies.(Please Note: Racks Purchased Separately are NOT Supplied with Vials.) Part of the CG-1869-01 Bill-Board set. The Bill-Board set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i.e. simultaneous silica gel chromatography).
Catalog No. 50-153-1269 Supplier Chemglass Life Sciences Supplier No. CG186914
Collection Vial Rack Only, 6-Place, Bill-Board. Have six wells to hold 25 x 52mm collection vials. Racks have four Stainless Steel pins that correspond to the holes in the top plate assemblies.(Please Note: Racks Purchased Separately are NOT Supplied with Vials.) Part of the CG-1869-01 Bill-Board set. The Bill-Board set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i.e. simultaneous silica gel chromatography).