Learn More
Glas Col LLC Control
SDP
Supplier:  Glas Col LLC 104A RL3612
Encompass_Preferred
Percentage timer for 6 place RJ (high temperature) or RX (low temperature) multi position combo mantle. 15A 120V
Catalog No. NC1479056