Learn More
Perkin Elmer US LLC 6-Port Valve, Aqueous Applications, for ASXPRESS PLUS
Catalog No. 502105317
Encompass_Preferred
The valve, manufactured from polyphenylene sulfide (PPS), contains six ports, each 1.52 mm in diameter. Labels for the ports are included.
Catalog No. 50-210-5317 Supplier Perkin Elmer US LLC Supplier No. N0777575
The valve, manufactured from polyphenylene sulfide (PPS), contains six ports, each 1.52 mm in diameter. Labels for the ports are included.