Learn More
Medchemexpress LLC 6PPD | 793-24-8 | 98.5% | 268.40 | 10 G
SDP
Catalog No. 5000453781
Encompass_Preferred
6PPD is a rubber antioxidant that scavenges ozone and forms nitro radicals. Exposure to 6PPD reduces the hatching rate, impairs spontaneous locomotor activity, shortens body length, and causes malformations in zebrafish embryos. It also induces oxidative stress in zebrafish embryos.
  • Rubber antioxidant
  • Scavenges ozone and forms nitro radicals
  • Causes concentration-dependent developmental toxicity, GH/IGF and HPT axis disruption, and oxidative stress in Danio rerio embryos/larvae
  • 96 h LC50 of 2.2 mg/L in zebrafish embryos
  • Reduced hatchability and autonomous movement in zebrafish embryos
Catalog No. 50-004-53781 Supplier Medchemexpress LLC Supplier No. HYW01168910G
Add to Cart
Add to Cart
6PPD is a rubber antioxidant that scavenges ozone and forms nitro radicals. Exposure to 6PPD reduces the hatching rate, impairs spontaneous locomotor activity, shortens body length, and causes malformations in zebrafish embryos. It also induces oxidative stress in zebrafish embryos.
  • Rubber antioxidant
  • Scavenges ozone and forms nitro radicals
  • Causes concentration-dependent developmental toxicity, GH/IGF and HPT axis disruption, and oxidative stress in Danio rerio embryos/larvae
  • 96 h LC50 of 2.2 mg/L in zebrafish embryos
  • Reduced hatchability and autonomous movement in zebrafish embryos