Learn More
Medchemexpress LLC 6ppd | 793-24-8 | 98.5% | 268.40 | 100 G
SDP
Catalog No. 5000452858
Encompass_Preferred
6PPD is a rubber antioxidant that functions by scavenging ozone and forming nitro radicals. Studies have shown that exposure to 6PPD can negatively impact zebrafish embryos, leading to reduced hatching rates, impaired spontaneous locomotor activity, shorter body length, and malformations. It also induces oxidative stress in these embryos.
  • Rubber antioxidant.
  • Scavenges ozone and forms nitro radicals.
Catalog No. 50-004-52858 Supplier Medchemexpress LLC Supplier No. HYW011689100G
6PPD is a rubber antioxidant that functions by scavenging ozone and forming nitro radicals. Studies have shown that exposure to 6PPD can negatively impact zebrafish embryos, leading to reduced hatching rates, impaired spontaneous locomotor activity, shorter body length, and malformations. It also induces oxidative stress in these embryos.
  • Rubber antioxidant.
  • Scavenges ozone and forms nitro radicals.