Learn More
Medchemexpress LLC 6PPD-Q-13C6 (6PPD-quinone-13C6) | 2754428-18-5 | C1213C6H22N2O2 | 100 UG
SDP
Catalog No. 5000374059
Encompass_Preferred
6PPD-quinone-13C6 is a 13C labeled compound, serving as both a tracer and an internal standard for quantitative analysis. It is utilized across various spectroscopic techniques, including Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), and Liquid Chromatography-Mass Spectrometry (LC-MS).
  • 13C labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS
Catalog No. 50-003-74059 Supplier Medchemexpress LLC Supplier No. HY152039S1100UG
6PPD-quinone-13C6 is a 13C labeled compound, serving as both a tracer and an internal standard for quantitative analysis. It is utilized across various spectroscopic techniques, including Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), and Liquid Chromatography-Mass Spectrometry (LC-MS).
  • 13C labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS