Learn More
Medchemexpress LLC 6PPD-quinone | 2754428-18-5 | 99.3% | 298.38 g/mol | C18H22N2O2 | 10 MG
SDP
Catalog No. 5000286538
Encompass_Preferred
6PPD-quinone is an oxidized derivative of the tire antioxidant 6-PPD, supplied as a high-purity research reagent for environmental and toxicological studies. It is commonly used as a reference standard for analytical method development, quantification in environmental matrices, and mechanistic investigations of toxicity.
  • High purity (99.3%).
  • CAS number 2754428-18-5 for unambiguous identification.
  • Suitable as an analytical reference standard for LC and GC methods.
  • Intended for environmental and toxicological research applications.
  • Available in milligram pack sizes for laboratory use.
Catalog No. 50-002-86538 Supplier Medchemexpress LLC Supplier No. HY15316910MG
6PPD-quinone is an oxidized derivative of the tire antioxidant 6-PPD, supplied as a high-purity research reagent for environmental and toxicological studies. It is commonly used as a reference standard for analytical method development, quantification in environmental matrices, and mechanistic investigations of toxicity.
  • High purity (99.3%).
  • CAS number 2754428-18-5 for unambiguous identification.
  • Suitable as an analytical reference standard for LC and GC methods.
  • Intended for environmental and toxicological research applications.
  • Available in milligram pack sizes for laboratory use.