Learn More
Medchemexpress LLC 6PPD-quinone-d5 | 2750119-14-1 | C18H17D5N2O2 | 1 MG
SDP
Catalog No. 5000373627
Encompass_Preferred
6PPD-quinone-d5 is the deuterium labeled 6PPD-quinone. It is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-73627 Supplier Medchemexpress LLC Supplier No. HY152039S1MG
6PPD-quinone-d5 is the deuterium labeled 6PPD-quinone. It is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.