Learn More
Medchemexpress LLC 6PPD-quinone-d5 | 2750119-14-1 | C18H17D5N2O2 | 5 MG
SDP
Catalog No. 5000373578
Encompass_Preferred
6PPD-quinone-d5 is a deuterium-labeled compound, specifically the deuterium-labeled form of 6PPD-quinone. It is utilized in drug development as a tracer and internal standard for quantitative analysis, primarily due to deuterium substitution's potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-73578 Supplier Medchemexpress LLC Supplier No. HY152039S5MG
6PPD-quinone-d5 is a deuterium-labeled compound, specifically the deuterium-labeled form of 6PPD-quinone. It is utilized in drug development as a tracer and internal standard for quantitative analysis, primarily due to deuterium substitution's potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS