Learn More
Medchemexpress LLC (6R,7S)-cefminox (sodium heptahydrate) | 92636-39-0 | 98.2% | 668.68 | C16H21N7O7S3.7H2O.Na | 100 MG
SDP
Catalog No. 5000239802
Encompass_Preferred
(6R,7S)-Cefminox sodium heptahydrate is an isomeric β-lactam cephalosporin antibiotic supplied for research use. It presents as a white to off-white solid and is available in both solid and solution formats suitable for biological and chemical studies.
  • Broad-spectrum antibacterial activity.
  • High reported purity (98.19%).
  • Available as solid and as DMSO solutions for reconstitution or ready-to-use.
  • Stable under recommended storage conditions (solid: 4°C; in solvent: -80°C for long-term storage).
  • Characterized molecular weight and formula for analytical reference.
Catalog No. 50-002-39802 Supplier Medchemexpress LLC Supplier No. HY107330100MG
(6R,7S)-Cefminox sodium heptahydrate is an isomeric β-lactam cephalosporin antibiotic supplied for research use. It presents as a white to off-white solid and is available in both solid and solution formats suitable for biological and chemical studies.
  • Broad-spectrum antibacterial activity.
  • High reported purity (98.19%).
  • Available as solid and as DMSO solutions for reconstitution or ready-to-use.
  • Stable under recommended storage conditions (solid: 4°C; in solvent: -80°C for long-term storage).
  • Characterized molecular weight and formula for analytical reference.