Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enzo Life Sciences (6R)-5,6,7,8-Tetrahydro-L-biopterin. dihydrochloride (25mg). CAS: 69056-38-8
SDP

Catalog No. 502009265
Encompass_Preferred

Cofactor of nitric oxide synthases (NOS). Alternative name: (6R)-BH4 . 2 HCl, [6R-[6R*(1R*,2S*)]]-2-Amino-6-(1,2-dihydroxypropyl)-5,6,7,8-tetrahydro-4(1H)-pteridinone . 2 HCl, Sapropterin dihydrochloride. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.

Catalog No. 50-200-9265 Supplier Enzo Life Sciences Supplier No. ALX440001M025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cofactor of nitric oxide synthases (NOS). Alternative name: (6R)-BH4 . 2 HCl, [6R-[6R*(1R*,2S*)]]-2-Amino-6-(1,2-dihydroxypropyl)-5,6,7,8-tetrahydro-4(1H)-pteridinone . 2 HCl, Sapropterin dihydrochloride. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.