Learn More
Cayman Chemical (6R,S)-5,6,7,8-Tetrahydrofolic Acid (hydrochloride), Purity: ≥95%, CAS: 150731-85-4, 10 mg
SDP
Catalog No. 502984603
Encompass_Preferred
A reduced form of folic acid that serves as a cofactor in methyltransferase reactions and is the major one-carbon carrier in one carbon metabolism; important for the synthesis of thymidine for incorporation into DNA or the synthesis of purines, as well as for the formation of methionine, which can be converted to the ubiquitous methyl donor, SAM
Catalog No. 50-298-4603 Supplier Cayman Chemical Supplier No. 2052610 MG
A reduced form of folic acid that serves as a cofactor in methyltransferase reactions and is the major one-carbon carrier in one carbon metabolism; important for the synthesis of thymidine for incorporation into DNA or the synthesis of purines, as well as for the formation of methionine, which can be converted to the ubiquitous methyl donor, SAM