Learn More
Cayman Chemical (6R,S)-5,10-Methenyl-5,6,7,8-tetrahydrofolic Acid (chloride), Purity: ≥95%, CAS: 65981-90-0, 10 mg
SDP
Catalog No. 502991699
Encompass_Preferred
An intermediate in the biosynthesis of thymidylate
Catalog No. 50-299-1699 Supplier Cayman Chemical Supplier No. 3133310 MG
An intermediate in the biosynthesis of thymidylate