Learn More
Cayman Chemical 6R5 6 7 8tetrahydroLBIo 10mg
SDP
Catalog No. 502895451
Encompass_Preferred
A cofactor for production of aromatic amino acids, neurotransmitters, and nitric oxide
Catalog No. 50-289-5451 Supplier Cayman Chemical Supplier No. 8188010 MG
A cofactor for production of aromatic amino acids, neurotransmitters, and nitric oxide