Learn More
Medchemexpress LLC (6S,12aR)-Tadalafil | 171596-28-4 | 99.5% | C22H19N3O4 | 5 MG
SDP
Catalog No. 5000432895
Encompass_Preferred
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is intended for research use only.
  • Highly potent PDE5 inhibitor
  • IC50 value of 5 nM
  • Exhibits blood pressure lowering activity
Catalog No. 50-004-32895 Supplier Medchemexpress LLC Supplier No. HY90009D5MG
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is intended for research use only.
  • Highly potent PDE5 inhibitor
  • IC50 value of 5 nM
  • Exhibits blood pressure lowering activity