Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (6S,12aR)-Tadalafil | 171596-28-4 | MFCD21363860 | 99.3% | 389.40 g·mol^-1 | C22H19N3O4 | 10 MG
SDP

Catalog No. 5000298679
Encompass_Preferred

(6S,12aR)-Tadalafil is a stereoisomeric impurity of tadalafil used as an analytical reference standard and research tool. It is a potent phosphodiesterase type 5 (PDE5) inhibitor (reported IC50 ≈ 5 nM) and is characterized for impurity profiling and pharmacological studies.

  • High chemical purity (reported 99.3%).
  • Well characterized: CAS 171596-28-4 and molecular formula C22H19N3O4.
  • Suitable as a reference standard for analytical method development and impurity profiling.
  • Useful for pharmacological research into PDE5 activity and blood-pressure effects.
  • Available in small weighed packs for laboratory use (e.g., 10 MG).

Catalog No. 50-002-98679 Supplier Medchemexpress LLC Supplier No. HY90009D10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(6S,12aR)-Tadalafil is a stereoisomeric impurity of tadalafil used as an analytical reference standard and research tool. It is a potent phosphodiesterase type 5 (PDE5) inhibitor (reported IC50 ≈ 5 nM) and is characterized for impurity profiling and pharmacological studies.

  • High chemical purity (reported 99.3%).
  • Well characterized: CAS 171596-28-4 and molecular formula C22H19N3O4.
  • Suitable as a reference standard for analytical method development and impurity profiling.
  • Useful for pharmacological research into PDE5 activity and blood-pressure effects.
  • Available in small weighed packs for laboratory use (e.g., 10 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.