Learn More
Medchemexpress LLC (6S,12aR)-Tadalafil (Tadalafil EP Impurity C) | 171596-28-4 | 99.5% | C22H19N3O4 | 25 MG
SDP
Catalog No. 5000357629
Encompass_Preferred
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM, known for its blood pressure lowering activity. This product is intended for research use only and presents as a white to off-white solid.
  • Highly potent PDE5 inhibitor with an IC50 value of 5 nM
  • Exhibits blood pressure lowering activity
  • Supplied as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Formulations available for in vivo studies
Catalog No. 50-003-57629 Supplier Medchemexpress LLC Supplier No. 25MGHY90009D
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM, known for its blood pressure lowering activity. This product is intended for research use only and presents as a white to off-white solid.
  • Highly potent PDE5 inhibitor with an IC50 value of 5 nM
  • Exhibits blood pressure lowering activity
  • Supplied as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Formulations available for in vivo studies