Learn More
Medchemexpress LLC (6S,12aR)-Tadalafil (Tadalafil EP Impurity C) | 171596-28-4 | 99.5% | C22H19N3O4 | 50 MG
SDP
Catalog No. 5000433083
Encompass_Preferred
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is intended for research use only and is not sold to patients.
  • Highly potent PDE5 inhibitor with an IC50 value of 5 nM.
  • Has blood pressure lowering activity.
  • Suitable for research applications only.
Catalog No. 50-004-33083 Supplier Medchemexpress LLC Supplier No. HY90009D50MG
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is intended for research use only and is not sold to patients.
  • Highly potent PDE5 inhibitor with an IC50 value of 5 nM.
  • Has blood pressure lowering activity.
  • Suitable for research applications only.