Learn More
Medchemexpress LLC (6S,12aR)-tadalafil (tadalafil EP impurity C; (6S,12aR)-IC-351) | 171596-28-4 | 99.5% | C22H19N3O4 | 1 MG
SDP
Catalog No. 5000433223
Encompass_Preferred
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is for research use only and not sold to patients.
  • Highly potent PDE5 inhibitor
  • IC50 value of 5 nM
  • Has blood pressure lowering activity
  • For research use only
  • Not sold to patients
Catalog No. 50-004-33223 Supplier Medchemexpress LLC Supplier No. HY90009D1MG
(6S,12aR)-Tadalafil (Tadalafil EP Impurity C) is a highly potent PDE5 inhibitor with an IC50 value of 5 nM and has blood pressure lowering activity. It is for research use only and not sold to patients.
  • Highly potent PDE5 inhibitor
  • IC50 value of 5 nM
  • Has blood pressure lowering activity
  • For research use only
  • Not sold to patients