Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (6S)-CP-470711 | 300552-09-4 | 98.8% | 358.44 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1646051MG

Encompass_Preferred

(6S)-CP-470711 (Compound 8) is an inhibitor for sorbitol dehydrogenase, inhibiting human and rat SDH with IC50 of 19 nM and 27 nM. It ameliorates Streptozotocin-induced diabetes in rat models. It is for research use only and not sold to patients.

  • Inhibitor for sorbitol dehydrogenase (SDH)
  • Inhibits human and rat SDH
  • Ameliorates Streptozotocin-induced diabetes in rat models
  • Appearance: Solid
  • Color: White to off-white
  • Molecular formula: C18H26N6O2

Catalog No. 50-003-92446


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.