Learn More
Medchemexpress LLC (6S)-CP-470711 | 300552-09-4 | 98.8% | 358.44 | 25 MG
SDP
Catalog No. 5000392277
Encompass_Preferred
(6S)-CP-470711 (Compound 8) is an inhibitor of sorbitol dehydrogenase (SDH), with IC50 values of 19 nM for human SDH and 27 nM for rat SDH. It has been shown to improve Streptozotocin-induced diabetes in rat models.
  • Inhibits human and rat sorbitol dehydrogenase (SDH) with IC50 values of 19 nM and 27 nM, respectively.
  • Ameliorates streptozotocin-induced diabetes in rat models.
  • Solid appearance, white to off-white color.
  • For research use only.
Catalog No. 50-003-92277 Supplier Medchemexpress LLC Supplier No. 25MGHY164605
(6S)-CP-470711 (Compound 8) is an inhibitor of sorbitol dehydrogenase (SDH), with IC50 values of 19 nM for human SDH and 27 nM for rat SDH. It has been shown to improve Streptozotocin-induced diabetes in rat models.
  • Inhibits human and rat sorbitol dehydrogenase (SDH) with IC50 values of 19 nM and 27 nM, respectively.
  • Ameliorates streptozotocin-induced diabetes in rat models.
  • Solid appearance, white to off-white color.
  • For research use only.