Learn More
Medchemexpress LLC (6S)-CP-470711 | 300552-09-4 | 98.8% | 358.44 | 5 MG
SDP
Catalog No. 5000392158
Encompass_Preferred
(6S)-CP-470711 (Compound 8) is an inhibitor for sorbitol dehydrogenase. It inhibits human and rat SDH with IC50 values of 19 nM and 27 nM, respectively. This compound ameliorates Streptozotocin-induced diabetes in rat models.
  • Inhibitor for sorbitol dehydrogenase (SDH).
  • Inhibits human and rat SDH with high potency (IC50 of 19 nM and 27 nM respectively).
  • Ameliorates Streptozotocin-induced diabetes in rat models.
  • Available in various pack sizes for research needs.
Catalog No. 50-003-92158 Supplier Medchemexpress LLC Supplier No. HY1646055MG
(6S)-CP-470711 (Compound 8) is an inhibitor for sorbitol dehydrogenase. It inhibits human and rat SDH with IC50 values of 19 nM and 27 nM, respectively. This compound ameliorates Streptozotocin-induced diabetes in rat models.
  • Inhibitor for sorbitol dehydrogenase (SDH).
  • Inhibits human and rat SDH with high potency (IC50 of 19 nM and 27 nM respectively).
  • Ameliorates Streptozotocin-induced diabetes in rat models.
  • Available in various pack sizes for research needs.