Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (6S)-CP-470711 | 300552-09-4 | 98.8% | 358.44 | C18H26N6O2 | 10 MG
SDP

Catalog No. 5000289559
Encompass_Preferred

(6S)-CP-470711 is a research-grade small-molecule inhibitor of sorbitol dehydrogenase (SDH). Supplied as a powder in small research pack sizes, it has low-nanomolar potency against human and rat SDH and is intended for laboratory research use.

  • Low-nanomolar SDH inhibitor (human IC50 19 nM; rat IC50 27 nM).
  • Powder form suitable for storage and handling.
  • Purity 98.75% by manufacturer specification.
  • Chemical formula C18H26N6O2; molecular weight 358.44.
  • Available in small pack sizes, including a 10 mg pack.
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for up to 6 months.

Catalog No. 50-002-89559 Supplier Medchemexpress LLC Supplier No. HY16460510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(6S)-CP-470711 is a research-grade small-molecule inhibitor of sorbitol dehydrogenase (SDH). Supplied as a powder in small research pack sizes, it has low-nanomolar potency against human and rat SDH and is intended for laboratory research use.

  • Low-nanomolar SDH inhibitor (human IC50 19 nM; rat IC50 27 nM).
  • Powder form suitable for storage and handling.
  • Purity 98.75% by manufacturer specification.
  • Chemical formula C18H26N6O2; molecular weight 358.44.
  • Available in small pack sizes, including a 10 mg pack.
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.