Learn More
Medchemexpress LLC (6S)-Tetrahydrofolic acid | 71963-69-4 | C19H23N7O6 | 50 MG
SDP
Catalog No. 5000355277
Encompass_Preferred
(6S)-Tetrahydrofolic acid is an endogenous metabolite that is significantly more active than its (6R) form. It promotes the binding of fluorodeoxyuridylate to thymidylate synthase and acts as a growth factor for P. cerevisiae. This compound also exhibits a low affinity and high dissociation rate for folate-binding protein.
  • Significantly more active than (6R) form
  • Promotes binding of fluorodeoxyuridylate to thymidylate synthase
  • Acts as a growth factor for P. cerevisiae
  • Exhibits low affinity and high dissociation rate for folate-binding protein
  • Endogenous metabolite with a molecular weight of 445.43
Catalog No. 50-003-55277 Supplier Medchemexpress LLC Supplier No. HY1452050MG
(6S)-Tetrahydrofolic acid is an endogenous metabolite that is significantly more active than its (6R) form. It promotes the binding of fluorodeoxyuridylate to thymidylate synthase and acts as a growth factor for P. cerevisiae. This compound also exhibits a low affinity and high dissociation rate for folate-binding protein.
  • Significantly more active than (6R) form
  • Promotes binding of fluorodeoxyuridylate to thymidylate synthase
  • Acts as a growth factor for P. cerevisiae
  • Exhibits low affinity and high dissociation rate for folate-binding protein
  • Endogenous metabolite with a molecular weight of 445.43