Learn More
Cayman Chemical 6ThIogunosIn5OtrIphosp 2.7mg
SDP
Catalog No. 502918242
Encompass_Preferred
An active metabolite of 6-thioguanine and azathioprine; binds to a variety of GTPases but selectively inhibits activation of Rac1 and Rac2 over Cdc42 in primary human CD4+ T cells stimulated with anti-CD3 and anti-CD28 antibodies; inhibits T cell receptor-stimulated proliferation of T cells and reduces CD28-dependent T cell survival in vitro in isolated mouse splenocytes; increases survival in a mouse model of heart transplantation at 0.5 mg/kg
Catalog No. 50-291-8242 Supplier Cayman Chemical Supplier No. 383902.7 MG
An active metabolite of 6-thioguanine and azathioprine; binds to a variety of GTPases but selectively inhibits activation of Rac1 and Rac2 over Cdc42 in primary human CD4+ T cells stimulated with anti-CD3 and anti-CD28 antibodies; inhibits T cell receptor-stimulated proliferation of T cells and reduces CD28-dependent T cell survival in vitro in isolated mouse splenocytes; increases survival in a mouse model of heart transplantation at 0.5 mg/kg