Learn More
Meiji Techno America Meiji Techno America Spare bulb 6V 30W halogen (for ML, MT, TC, IM Halogen models)
Catalog No. NC9446156
Encompass_Preferred
Spare bulb 6V 30W halogen (for ML, MT, TC, IM Halogen models)
Catalog No. NC9446156 Supplier Meiji Techno America Supplier No. MA326
Spare bulb 6V 30W halogen (for ML, MT, TC, IM Halogen models)