Learn More
Caplugs 6WELL PLTS W/LID STERILE UNTRE
Catalog No. NC2459870
Encompass_Preferred
Caplugs Evergreen 6-well Plates are gamma-irradiated for sterility and available in both sterile and treated versions.Distinct alpha-numeric well designationsWell volume: 17.7 mLLid included with plate-Items are not available for shipment to Mexico
Catalog No. NC2459870 Supplier Caplugs Supplier No. 222804801F
Caplugs Evergreen 6-well Plates are gamma-irradiated for sterility and available in both sterile and treated versions.Distinct alpha-numeric well designationsWell volume: 17.7 mLLid included with plate-Items are not available for shipment to Mexico