Learn More
Sarstedt Inc Cell Culture Plate, 6 well, surface: Suspension, flat base
Catalog No. NC2029375 Shop All Sarstedt Inc. Products
Encompass_Preferred
Cell culture plate, 6 well, material: PS, surface: Suspension, for suspension cells, coding colour: green, flat base, TC Tested, 1 piece(s)/blister.
Catalog No. NC2029375 Supplier Sarstedt Inc Supplier No. 83.3920.500
Cell culture plate, 6 well, material: PS, surface: Suspension, for suspension cells, coding colour: green, flat base, TC Tested, 1 piece(s)/blister.