Learn More
Thomas Scientific Bag, Biohazard Specimen Transport, 6" x 9", Glue Seal with Document Pouch, 100/Pack, 10 Packs/Unit
SDP
Catalog No. 501460360
Encompass_Preferred
These bags are designed for the safe transportation of specimens and documentation. Each bag is printed with the biohazard symbol and offers checkmark boxes to indicate special handling instructions. 2 mil gauge polyethylene (PE) Attached document compartment Zipper closure allows repeated access to specimen Imprinted with biohazard symbol Complies with OSHA and NCCLS guidelines
This item is not returnable. View return policy
Catalog No. 50-146-0360 Supplier Thomas Scientific Supplier No. 1202H00
These bags are designed for the safe transportation of specimens and documentation. Each bag is printed with the biohazard symbol and offers checkmark boxes to indicate special handling instructions. 2 mil gauge polyethylene (PE) Attached document compartment Zipper closure allows repeated access to specimen Imprinted with biohazard symbol Complies with OSHA and NCCLS guidelines