Learn More
Gold Biotechnology Inc 6X GREEN LOADING DYE 5ML
SDP
Catalog No. NC2396777
Encompass
GoldBio's 6x Green DNA Loading Dye is a pre-mixed buffer for tracking DNA samples during the electrophoresis on agarose or polyacrylamide gels. It contains two dyes (Xylene cyanol FF and Orange G) for tracking the DNA migration. The Xylene cyanol FF and Orange G migrate at approximately 4,000 bp and 50 bp on a standard 1% TAE agarose gel respectively (800 bp and 30 bp on a 2% TAE agarose gel respectively). The included glycerol keeps the DNA at the bottom of the well and the presence of EDTA chelates divalent metal ions to prevent the process of metal-dependent nuclease.
Catalog No. NC2396777 Supplier Gold Biotechnology Inc Supplier No. L00155ML
GoldBio's 6x Green DNA Loading Dye is a pre-mixed buffer for tracking DNA samples during the electrophoresis on agarose or polyacrylamide gels. It contains two dyes (Xylene cyanol FF and Orange G) for tracking the DNA migration. The Xylene cyanol FF and Orange G migrate at approximately 4,000 bp and 50 bp on a standard 1% TAE agarose gel respectively (800 bp and 30 bp on a 2% TAE agarose gel respectively). The included glycerol keeps the DNA at the bottom of the well and the presence of EDTA chelates divalent metal ions to prevent the process of metal-dependent nuclease.