Learn More
IBI SCIENTIFIC 3P 6x Gel Electrophoresis Loading Dye, 5 mL
Catalog No. NC0751928
Encompass_Preferred
IBI 6X Loading Dye contains 15% ficoll in dyes that make estimating sample migration easy and reliable.
Catalog No. NC0751928 Supplier IBI SCIENTIFIC 3P Supplier No. IB01010
IBI 6X Loading Dye contains 15% ficoll in dyes that make estimating sample migration easy and reliable.