Learn More
ANALYTICAL CONTROL SYSTEMS INC Variant Hemoglobin Control, 6 vials, 0.5 mL each, Lyophilized powder, Hemoglobin A 40%, Hemoglobin F 25%, Hemoglobin S 20%, Hemoglobin C 15%, Reconstituted Stability 10 days Refrigerated, Unreconstituted Stability 2 years Refrigerated
SDP
Catalog No. NC1304293
Encompass
Variant Hemoglobin Control, 6 vials, 0.5 mL each, Lyophilized powder, Hemoglobin A 40%, Hemoglobin F 25%, Hemoglobin S 20%, Hemoglobin C 15%, Reconstituted Stability 10 days Refrigerated, Unreconstituted Stability 2 years Refrigerated
Catalog No. NC1304293 Supplier ANALYTICAL CONTROL SYSTEMS INC Supplier No. HC102
Variant Hemoglobin Control, 6 vials, 0.5 mL each, Lyophilized powder, Hemoglobin A 40%, Hemoglobin F 25%, Hemoglobin S 20%, Hemoglobin C 15%, Reconstituted Stability 10 days Refrigerated, Unreconstituted Stability 2 years Refrigerated