Learn More
ILEX LIFE SCIENCES LLC 6 x 30 mm Ossiform P3D Scaffolds, suitable for 6-well plates - gyroid structure, 600-800 micrometers porosity, bone-like 3D cell culture systems
SDP
Catalog No. NC2209133
Encompass_Preferred
Ossiform P3D Scaffolds are xeno-free, bioceramic scaffolds which provide a bone-like niche for in vitro and in vivo studies such as disease modeling, drug screening and bioengineering. The 3D printed scaffolds are developed to resemble native bone with respect to both physical and structural properties, such as high contents of β-tricalcium phosphate (β-TCP) and stiffness, as well as macro- and micro-porosities. The P3D Scaffolds are an easy plug-and-play solution for 3D cell culturing of bone harboring or bone related cell types. P3D Scaffolds are compatible with most common laboratory techniques and do not require major alterations of preexisting protocols and workflows. P3D Scaffolds provide you with a clinically relevant cell culture system of natural materials and customized structures which allow you to create predictive research models of human physiology and pathology.
This item is not returnable. View return policy
Catalog No. NC2209133 Supplier ILEX LIFE SCIENCES LLC Supplier No. DRL06GY2
Ossiform P3D Scaffolds are xeno-free, bioceramic scaffolds which provide a bone-like niche for in vitro and in vivo studies such as disease modeling, drug screening and bioengineering. The 3D printed scaffolds are developed to resemble native bone with respect to both physical and structural properties, such as high contents of β-tricalcium phosphate (β-TCP) and stiffness, as well as macro- and micro-porosities. The P3D Scaffolds are an easy plug-and-play solution for 3D cell culturing of bone harboring or bone related cell types. P3D Scaffolds are compatible with most common laboratory techniques and do not require major alterations of preexisting protocols and workflows. P3D Scaffolds provide you with a clinically relevant cell culture system of natural materials and customized structures which allow you to create predictive research models of human physiology and pathology.