Learn More
Elkay Plastics Company Bag Clear Slide Seal 3 mil 6 X 6 250 per Case
Catalog No. NC9583936
Encompass_Preferred
LK Slide Seal Bags are excellent for storing, displaying and protecting all types of products, especially those that need to be accessed multiple times. The slide seal closure is easy to use, even with gloved hands. The bags meet FDA requirements for use with food products.
Catalog No. NC9583936 Supplier Elkay Plastics Company Supplier No. FSL30606
LK Slide Seal Bags are excellent for storing, displaying and protecting all types of products, especially those that need to be accessed multiple times. The slide seal closure is easy to use, even with gloved hands. The bags meet FDA requirements for use with food products.