Learn More
Uline Reclosable Static Shielding Bag, Pack of 100
Catalog No. NC2380141
Encompass_Preferred
6""""x6"""" polyethylene antistatic bags. Secure static-sensitive items quickly with double-track zipper closure. Ideal for intercompany transfers, metal """"Faraday cage"""" layer shields products from electric energy inside and prevents static buildup, 3 mil transparent metallic film, 4-layer protection guards against charges inside and out.
Catalog No. NC2380141 Supplier Uline Supplier No. S7624
6""""x6"""" polyethylene antistatic bags. Secure static-sensitive items quickly with double-track zipper closure. Ideal for intercompany transfers, metal """"Faraday cage"""" layer shields products from electric energy inside and prevents static buildup, 3 mil transparent metallic film, 4-layer protection guards against charges inside and out.