Learn More
Uline 6 x 8"" 4 Mil Reclosable Bags
Catalog No. NC1714219
Encompass_Preferred
Heavyweight poly bags provide total protection from oil, dust and moisture. ¿Keep items clean and ready to use. ¿100% virgin polyethylene film. FDA and USDA compliant. ¿Combine sizes for volume discounts. MATERIAL: LDPE (Low Density Polyethylene)
Catalog No. NC1714219 Supplier Uline Supplier No. S1711
Heavyweight poly bags provide total protection from oil, dust and moisture. ¿Keep items clean and ready to use. ¿100% virgin polyethylene film. FDA and USDA compliant. ¿Combine sizes for volume discounts. MATERIAL: LDPE (Low Density Polyethylene)