Learn More
Elkay Plastics Company Bag Specimen Transport Zip Closure 3 wall Red Opaque with Biohazard print and an Outside Pouch 2 mil 6 X 9 1,000 per Case
Catalog No. NC0416838
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags provide an excellent means for transferring specimens in hospital and medical environments. Three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer. Bags are inner packed in 100s.
Catalog No. NC0416838 Supplier Elkay Plastics Company Supplier No. LAB20609ROP
Add to Cart
Add to Cart
LKs Lab-Loc Reclosable Zipper Specimen Bags provide an excellent means for transferring specimens in hospital and medical environments. Three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer. Bags are inner packed in 100s.