Learn More
Ubpbio N-terminally 6xHis-tagged recombinant human UbE2R1, a ubiquitin conjugating enzyme catalyzing formation of K48 polyubiquitin chains
Catalog No. 502339338
Encompass_Preferred
Ube2R1 (UbcH3) is an E2 enzyme, which is part of the E1, E2, and E3 cascade responsible for ubiquitination of protein substrates. Ube2R1 was first discovered during screening for cell cycle regulatory mutations that occur in yeast. While Ube2R1 is able to create diubiquitin chains by itself, it works far more efficiently and effectively with the E3 ligase Skp1 (S- phase kinase- associated protein 1)/cullin/F-box, also known as SCF, to promote substrate polyubiquitination.
Catalog No. 50-233-9338 Supplier Ubpbio Supplier No. C3500
Ube2R1 (UbcH3) is an E2 enzyme, which is part of the E1, E2, and E3 cascade responsible for ubiquitination of protein substrates. Ube2R1 was first discovered during screening for cell cycle regulatory mutations that occur in yeast. While Ube2R1 is able to create diubiquitin chains by itself, it works far more efficiently and effectively with the E3 ligase Skp1 (S- phase kinase- associated protein 1)/cullin/F-box, also known as SCF, to promote substrate polyubiquitination.