Learn More
Sklar Instruments Scissors, Multi-Cut Utility Scissors, Sklar lack, 7-1/2 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA.
SDP
Catalog No. 501093313 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Multi-Cut Utility Scissors, Sklar, Black, 7-1/2 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA. Sklar Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.
Catalog No. 50-109-3313 Supplier Sklar Instruments Supplier No. 111281
Scissors, Multi-Cut Utility Scissors, Sklar, Black, 7-1/2 in., Blunt/Blunt, Angled, Serrated, Stainless Steel / Plastic, EA. Sklar Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.