Learn More
Medchemexpress LLC 7-(1,3-dioxan-2-yl)-8-hydroxy-1,2,3,4-tetrahydro-5H-chromeno[3,4-c]pyridin-5-one | 1607803-67-7 | MFCD30742940 | 99.4% | 303.31 g·mol⁻¹ | C16H17NO5 | 5 MG
SDP
Catalog No. 5000239861
Encompass_Preferred
B I09 is a small-molecule research inhibitor targeting the IRE-1 RNase domain. Provided as a solid with high analytical purity, it is intended for in vitro biochemical and cell-based studies probing unfolded protein response signaling. The compound has a reported IC50 of 1,230 nM against IRE-1 RNase. Store at 4°C and protect from light. Not for human or clinical use.
  • Inhibits IRE-1 RNase activity with a reported IC50 of 1,230 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Solid form enables precise weighing and formulation.
  • Available in small research quantities for screening and validation.
  • Accompanied by analytical documentation including datasheet, COA, and SDS.
Catalog No. 50-002-39861 Supplier Medchemexpress LLC Supplier No. HY1074005MG
B I09 is a small-molecule research inhibitor targeting the IRE-1 RNase domain. Provided as a solid with high analytical purity, it is intended for in vitro biochemical and cell-based studies probing unfolded protein response signaling. The compound has a reported IC50 of 1,230 nM against IRE-1 RNase. Store at 4°C and protect from light. Not for human or clinical use.
  • Inhibits IRE-1 RNase activity with a reported IC50 of 1,230 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Solid form enables precise weighing and formulation.
  • Available in small research quantities for screening and validation.
  • Accompanied by analytical documentation including datasheet, COA, and SDS.