Learn More
Sklar Instruments Forceps, Cushing Forceps Bayonet, Sklar 7-1/4 in., Dressing Forceps, Straight, Serrated, Bayonet, EA.
SDP
Catalog No. 501098160 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Cushing Forceps Bayonet, Sklar, 7-1/4 in., Dressing Forceps, Straight, Serrated, Bayonet, EA. Sklar Cushing Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. These forceps may also be known as Cushing-Taylor Forceps. This product is has a bayonet handle with straight jaws. The tips are serrated and the overall length is 7-1/4 inches.
Catalog No. 50-109-8160 Supplier Sklar Instruments Supplier No. 226970
Forceps, Cushing Forceps Bayonet, Sklar, 7-1/4 in., Dressing Forceps, Straight, Serrated, Bayonet, EA. Sklar Cushing Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. These forceps may also be known as Cushing-Taylor Forceps. This product is has a bayonet handle with straight jaws. The tips are serrated and the overall length is 7-1/4 inches.