Learn More
Sklar Instruments Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar 7-1/4 in., Straight, Serrated, Stainless Steel, Disposable, CS of 25.
SDP
Catalog No. 501094372 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar, 7 1/4 in., Straight, Serrated, Stainless Steel, Disposable, CS of 25. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
Catalog No. 50-109-4372 Supplier Sklar Instruments Supplier No. 963006
Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar, 7 1/4 in., Straight, Serrated, Stainless Steel, Disposable, CS of 25. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.